Learn More
Norgen Biotek Corp Plasma/Serum Cell-Free Circulating DNA Purification Midi Kit
Catalog No. NC0873156
Encompass_Preferred
This kit provides a fast, reliable and convenient method to purify and concentrate using a two column method for the isolation of high quality, high purity and inhibitor-free cell-free circulating DNA (cfc-DNA) from fresh or frozen plasma/serum sample volumes ranging from 1 mL - 4 mL.The first column will handle the large volume input of bodily fluids that is followed by a concentration on a mini column for a final elution of 25 µL to 50 µL. All components for the purification & concentration are provided in one convenient & fast kit for the easy processing of large input volumes of bodily fluids.The purified plasma/serum cfc-DNA is fully compatible with all downstream applications including PCR, qPCR, methylation-sensitive PCR and Southern Blot analysis, microarrays and NGS, for various analyses: Fetal DNA screening, genotyping, and other oncology and autoimmune diseases research.
Catalog No. NC0873156 Supplier Norgen Biotek Corp Supplier No. 55600
This kit provides a fast, reliable and convenient method to purify and concentrate using a two column method for the isolation of high quality, high purity and inhibitor-free cell-free circulating DNA (cfc-DNA) from fresh or frozen plasma/serum sample volumes ranging from 1 mL - 4 mL.The first column will handle the large volume input of bodily fluids that is followed by a concentration on a mini column for a final elution of 25 µL to 50 µL. All components for the purification & concentration are provided in one convenient & fast kit for the easy processing of large input volumes of bodily fluids.The purified plasma/serum cfc-DNA is fully compatible with all downstream applications including PCR, qPCR, methylation-sensitive PCR and Southern Blot analysis, microarrays and NGS, for various analyses: Fetal DNA screening, genotyping, and other oncology and autoimmune diseases research.