Learn More
BEZWADA BIOMEDICAL LLC Poly(L-lactide-co-D,L-Lactide), L-lactide:D,L-lactide 70:30, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904469
Encompass_Preferred
PLA has a significantly higher crystallinity and degradation time than poly(D,L-lactide). This is attributed to the presence of all of the methyl units arranged on one side of the polymer chain sequence which results in dense packaging. Hence the presence of D,L-PLA repeat units in the L-PLA homopolymer repeat units reduces the crystallinity and enables faster hydrolytic degradation rate of L-PLA. The polymers are used in medical device, drug delivery and 3D printing.
Catalog No. 50-190-4469 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 3473
PLA has a significantly higher crystallinity and degradation time than poly(D,L-lactide). This is attributed to the presence of all of the methyl units arranged on one side of the polymer chain sequence which results in dense packaging. Hence the presence of D,L-PLA repeat units in the L-PLA homopolymer repeat units reduces the crystallinity and enables faster hydrolytic degradation rate of L-PLA. The polymers are used in medical device, drug delivery and 3D printing.