Learn More
BEZWADA BIOMEDICAL LLC Poly(L-lactide-co-glycolide), L-lactide:glycolide 85:15,viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904464
Encompass_Preferred
The complete absorption of L(-)-PLA requires more than 2 years and is much slower than that of DL-PLA. In order to decrease the absorption times of implantable devices incorporating L(-)-lactide, copolymers of L(-)-lactide and dl-lactide with glycolide have been prepared. The polymers are used in medical devices
Catalog No. 50-190-4464 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 3281
The complete absorption of L(-)-PLA requires more than 2 years and is much slower than that of DL-PLA. In order to decrease the absorption times of implantable devices incorporating L(-)-lactide, copolymers of L(-)-lactide and dl-lactide with glycolide have been prepared. The polymers are used in medical devices