Learn More
Sino Biological Recombinant Chinese hamster Phospholipase A2 group XV/PLA2G15 Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 5A7663A08C1
Encompass_Preferred
A DNA sequence encoding the chinese hamster PLA2G15 (G3HKV9-1) (Val34-Pro412) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6515