Learn More
Aviva Systems Biology Corp PLA2G4B Antibody - N-terminal region (ARP38556_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38556_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLA2G4B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLA2G4B. Alternate names: FLJ42498, HsT16992, cPLA2-beta, PLA2G4BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56702