Learn More
Aviva Systems Biology Corp PLA2G4E Antibody - C-terminal region (ARP53548_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53548_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLA2G4E. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the c terminal region of human PLA2G4E. Alternate names: FLJ45651, MGC126633, MGC126661ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-63042