Learn More
Aviva Systems Biology Corp PLA2G5 Antibody - middle region (ARP41429_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41429_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLA2G5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PLA2G5. Alternate names: DKFZp686C2294, GV-PLA2, MGC46205, PLA2-10, hVPLA(2), FRFBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-57848