Learn More
Aviva Systems Biology Corp PLAC9 Antibody - middle region (ARP47962_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLAC9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PLAC9. Alternate names: MGC104710ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-59688