Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVOS™ Light Cube, Cy5.5 2.0

Catalog No. 12563690
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-563-690 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-563-690 Supplier Invitrogen™ Supplier No. AMEP4973
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Second-generation EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.

TRUSTED_SUSTAINABILITY

Specifications

Type LED Light Cube
Product Line EVOS
Excitation/Emission Ex: 655/40 nm Em: 725/40
For Use With (Equipment) Countess™ III FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Includes Condensing Optics, Hard-coated Filters
Quantity 1 each
Are EVOS 2.0 Light Cubes compatible with the Countess II FL?

Yes. All our EVOS Light Cubes can be used with the Countess II FL.

Are EVOS 2.0 Light Cubes compatible with older EVOS instruments?

Yes. Our EVOS 2.0 Light cubes can be used in all EVOS instruments with EVOS light cube functionality.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.