Learn More
Bioss Placental alkaline phosphatase
SDP
Supplier:  Bioss BSM61630R
Encompass_Preferred
Placental alkaline phosphatase (PLAP) Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IHC-P IHC-F IF(ICC) IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-67623