Learn More
Medchemexpress LLC Placental growth factor-2, human (mFc fusion) | 97.5% | 20 UG
SDP
Catalog No. 5000291214
Encompass_Preferred
Recombinant human placental growth factor-2 (PLGF-2) produced in HEK293 cells with a C-terminal mFc fusion; provided for research on angiogenesis, endothelial cell biology, and receptor interactions.
  • Produced in HEK293 cells to support mammalian post-translational modifications.
  • Contains a C-terminal mFc fusion tag for increased stability and detection.
  • Promotes angiogenesis and endothelial cell proliferation via FLT1/VEGFR-1 and NRP1/NRP2 binding.
  • Suitable for receptor binding, signaling, and cell-based assays.
  • High purity (97.5%) and supplied in small research pack sizes.
Catalog No. 50-002-91214 Supplier Medchemexpress LLC Supplier No. HYP7462620UG
Recombinant human placental growth factor-2 (PLGF-2) produced in HEK293 cells with a C-terminal mFc fusion; provided for research on angiogenesis, endothelial cell biology, and receptor interactions.
  • Produced in HEK293 cells to support mammalian post-translational modifications.
  • Contains a C-terminal mFc fusion tag for increased stability and detection.
  • Promotes angiogenesis and endothelial cell proliferation via FLT1/VEGFR-1 and NRP1/NRP2 binding.
  • Suitable for receptor binding, signaling, and cell-based assays.
  • High purity (97.5%) and supplied in small research pack sizes.