Learn More
Creative Biomart Placental Growth Factor (PGF) Protein
Catalog No. 502084328
Encompass_Preferred
PGF1 Mouse Recombinant produced in Sf9 Baculovirus cells is a single, glycosylated polypeptide chain containing 138 amino acids (27-158a.a.) and having a molecular mass of 15.9kDa (Molecular size on SDS-PAGE will appear at approximately 18-28kDa). PGF1 is expressed with a 6 amino acid His tag at C-Terminus and purified by proprietary chromatographic techniques. Placental growth factor is a protein that in humans is encoded by the PGF gene.
Catalog No. 50-208-4328 Supplier Creative Biomart Supplier No. THP0277
PGF1 Mouse Recombinant produced in Sf9 Baculovirus cells is a single, glycosylated polypeptide chain containing 138 amino acids (27-158a.a.) and having a molecular mass of 15.9kDa (Molecular size on SDS-PAGE will appear at approximately 18-28kDa). PGF1 is expressed with a 6 amino acid His tag at C-Terminus and purified by proprietary chromatographic techniques. Placental growth factor is a protein that in humans is encoded by the PGF gene.