Learn More
Cayman Chemical Pladienolide B 100UG
SDP
Catalog No. 502968091
Encompass_Preferred
A macrocyclic lactone that selectively binds SF3b and inhibits mRNA splicing; potently blocks the growth of proliferating cells with mean IC50 values of 1.6 nM for six gastric cancer cell lines
Catalog No. 50-296-8091 Supplier Cayman Chemical Supplier No. 16538100 UG
A macrocyclic lactone that selectively binds SF3b and inhibits mRNA splicing; potently blocks the growth of proliferating cells with mean IC50 values of 1.6 nM for six gastric cancer cell lines