Learn More
Aviva Systems Biology Corp PLAG1 Antibody - C-terminal region (ARP38178_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38178_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLAG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PLAG1. Alternate names: SGPA, ZNF912ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56554