Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextromethorphan Hydrobromide, Monohydrate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602107 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-107 50 kg Amber Glass Bottle
18-602-101 10 g Amber Glass Bottle
18-602-102 25 g Amber Glass Bottle
18-602-103 100 g Amber Glass Bottle
18-602-106 500 g Amber Glass Bottle
18-602-104 1 kg Amber Glass Bottle
18-602-105 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-602-107 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DE13950KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

DE139, 6700-34-1, C18H25NO·HBr·H2O

Spectrum™ Chemical Dextromethorphan Hydrobromide, Monohydrate, USP is as a cough suppressant, for the temporary relief of cough. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6700-34-1
Molecular Formula C18H28BrNO2
Molecular Weight (g/mol) 370.33
InChI Key STTADZBLEUMJRG-IKNOHUQMSA-N
IUPAC Name hydrogen (1S,9S,10S)-4-methoxy-17-methyl-17-azatetracyclo[7.5.3.01,10.02,7]heptadeca-2,4,6-triene hydrate bromide
SMILES [H+].O.[Br-].COC1=CC=C2C[C@H]3[C@H]4CCCC[C@@]4(CCN3C)C2=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 50 kg
UN Number UN3077
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Dextromethorphan Hydrobromide, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.