Learn More
Grainger Plain Ball SS 3/32 in Rope dia.
Catalog No. 502767069
Encompass_Preferred
The Grainger 16X617 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-scratching fibers for cleaning and scouring. This 6 inch by 9 inch hand pad has a medium duty abrasiveness that is suitable for scrubbing pots, pans, dishes, floors, walls, and other surfaces. It effectively removes grease, dirt, food residue, and more without damaging the underlying surface. The pad is disposable and ideal for kitchen, bathroom, garage, and workshop cleaning tasks.
Catalog No. 50-276-7069 Supplier Grainger Supplier No. 16X617
The Grainger 16X617 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-scratching fibers for cleaning and scouring. This 6 inch by 9 inch hand pad has a medium duty abrasiveness that is suitable for scrubbing pots, pans, dishes, floors, walls, and other surfaces. It effectively removes grease, dirt, food residue, and more without damaging the underlying surface. The pad is disposable and ideal for kitchen, bathroom, garage, and workshop cleaning tasks.