Learn More
Foxx Life Sciences Abdos Desiccator with a clear Polycarbonate (PC) top and a white Polypropylene (PP) Plate (Diameter 300mm) 1/EA
Catalog No. NC1818241
Encompass_Preferred
Desiccators are enclosures containing desiccants for preserving moisture sensitive products. These desiccators are molded with high-quality plastics which are light and easy to handle. The Polycarbonate (PC) top is clear for easy viewing while the bottom part of the desiccators are white Polypropylene (PP) and come with a thick white perforated disc. Silicone O-ring assures tight seal down to 0C without using grease. Use with disposable or reusable desiccantFoxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.
Catalog No. NC1818241 Supplier Foxx Life Sciences Supplier No. E11604
Desiccators are enclosures containing desiccants for preserving moisture sensitive products. These desiccators are molded with high-quality plastics which are light and easy to handle. The Polycarbonate (PC) top is clear for easy viewing while the bottom part of the desiccators are white Polypropylene (PP) and come with a thick white perforated disc. Silicone O-ring assures tight seal down to 0C without using grease. Use with disposable or reusable desiccantFoxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.