Learn More
Andwin Scientific Microscope Glass Slides-Esco: Microscope Slide, Plain End: 1.2 mm Thick, 25x75, 144/pk
SDP
Catalog No. 50949344
Encompass_Preferred
Histology & Light Microscopy: Plastic, Glass, Quartz, Permanox, Polystyrene, PTFE Coated, Poly-L-Lysine, & Silane Treated Slides: Microscope Glass Slides-Esco: Microscope Slide, Plain End: 1.2 mm Thick, 25x75, 144/pk
Catalog No. 50-949-344 Supplier Andwin Scientific Supplier No. 7188001
Histology & Light Microscopy: Plastic, Glass, Quartz, Permanox, Polystyrene, PTFE Coated, Poly-L-Lysine, & Silane Treated Slides: Microscope Glass Slides-Esco: Microscope Slide, Plain End: 1.2 mm Thick, 25x75, 144/pk