Learn More
Formumax Scientific Inc PLAIN HSPC/CHOL/MPEG2000-DSPE
Catalog No. NC1946963
Encompass_Preferred
Plain HSPC/CHOL/mPEG2000-DSPE Liposomes (50:45:5 mol/mol, 100 nm)
Catalog No. NC1946963 Supplier Formumax Scientific Inc Supplier No. F10204
Plain HSPC/CHOL/mPEG2000-DSPE Liposomes (50:45:5 mol/mol, 100 nm)