Learn More
Bellco Glass, Inc. Plain Slide,25x75mm
SDP
Supplier:  Bellco Glass, Inc. 563812013
Encompass_Preferred
Pre-cleaned plain microscope slides with square corners and ground edges. Ideal for fluorescence and confocal microscopy. MADE IN THE USA Thickness: 1.10mm -1.30mm Size: 25x75mm
Catalog No. 50-194-5828