Learn More
Bellco Glass, Inc. Plain Slide,25x75mm
SDP
Catalog No. 501945827
Encompass_Preferred
Pre-cleaned plain microscope slides with square corners and ground edges. Ideal for fluorescence and confocal microscopy. MADE IN THE USA Thickness: 0.9mm -1.09mm Size: 25x75mm
Catalog No. 50-194-5827 Supplier Bellco Glass, Inc. Supplier No. 563811013
Pre-cleaned plain microscope slides with square corners and ground edges. Ideal for fluorescence and confocal microscopy. MADE IN THE USA Thickness: 0.9mm -1.09mm Size: 25x75mm