Learn More
Fisherbrand™ Plan Achromatic Objective 100X
GREENER_CHOICE
Catalog No. 03000117 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Lens
Catalog No. Type
03-000-117 Lens
1 options
Catalog No. 03-000-117
Add to Cart
Add to Cart
Plan achromatic objective.
  • Plan achromatic 100x objective
  • N.A: 1.25
  • W.D: 0.18 mm
  • F.N: 22
  • Coverslip Thickness: 0.17
  • Immersion Medium: Oil
  • Spring:  Yes
  • Illumination techniques:  BF  FL
Specifications
Type Lens
For Use With (Equipment) Upright Microscope
Immersion Type Oil
Magnification Power 100X
Numerical Aperture 1.25