Learn More
Electron Microscopy Sciences Tripod Polisher Planarizing Tool
SDP
Catalog No. 50980873
Encompass
The Planarizing Tool (#50116-40): is used to lap the three delrin feet to be coplanar in preparation for preparing samples with Parallel Polishing Mount. - For use with Tripod Polisher Model 590.
Catalog No. 50-980-873 Supplier Electron Microscopy Sciences Supplier No. 5011640
The Planarizing Tool (#50116-40): is used to lap the three delrin feet to be coplanar in preparation for preparing samples with Parallel Polishing Mount. - For use with Tripod Polisher Model 590.