Learn More
Grainger Planer Corded 120V AC
Catalog No. 502753425
Encompass_Preferred
The Makita 6-1/4 in. Corded Hand Planer (14F144) is a powerful tool for fast stock removal and smooth finishes. It features a 15 Amp motor delivering up to 16,000 RPM, a maximum cutting depth of 1/8 in., and can plane boards up to 6-1/4 in. wide. It has dual blade cutting heads, a lock-on button for continuous operation, a kickstand, and a dust bag. Ideal for professional contractors, carpenters, woodworkers, and DIYers, this planer ensures efficient planning of wood boards and surfaces.
Catalog No. 50-275-3425 Supplier Grainger Supplier No. 14F144
The Makita 6-1/4 in. Corded Hand Planer (14F144) is a powerful tool for fast stock removal and smooth finishes. It features a 15 Amp motor delivering up to 16,000 RPM, a maximum cutting depth of 1/8 in., and can plane boards up to 6-1/4 in. wide. It has dual blade cutting heads, a lock-on button for continuous operation, a kickstand, and a dust bag. Ideal for professional contractors, carpenters, woodworkers, and DIYers, this planer ensures efficient planning of wood boards and surfaces.