Learn More
Research Products International Corp Planet-Safe Petri Dish 375/cs
SDP
Catalog No. 502388372
Encompass
Planet-Safe Petri Dishes are made from polylactic acid (PLA), a biodegradable and compostable resin derived from a variety of non-GMO renewable resources. Ideal for growing bacteria, yeasts and molds. Offer excellent optical transparency and enable microscopic examination of cells while resisting organic solvents such as alcohols, ketones and hydrocarbons.
Engineered with a lower carbon footprint and 100% renewable resources to meet the needs of laboratories focusing on more Eco-friendly and sustainable practices. PLA bioresin is substantially more hydrophilic than petrochemical resins
such as polystryrene. This beneficial property allows a variety of anchorage-dependent eukaryotic cell species to grow in liquid culture on the petri dish surface without adding coatings such as ECM (extracellular matrices), proteines, collagen, fibronectin, poly-D-lysine, or specialty plasma gas treatments.
Catalog No. 50-238-8372 Supplier Research Products International Corp Supplier No. 160269
Planet-Safe Petri Dishes are made from polylactic acid (PLA), a biodegradable and compostable resin derived from a variety of non-GMO renewable resources. Ideal for growing bacteria, yeasts and molds. Offer excellent optical transparency and enable microscopic examination of cells while resisting organic solvents such as alcohols, ketones and hydrocarbons.
Engineered with a lower carbon footprint and 100% renewable resources to meet the needs of laboratories focusing on more Eco-friendly and sustainable practices. PLA bioresin is substantially more hydrophilic than petrochemical resins
such as polystryrene. This beneficial property allows a variety of anchorage-dependent eukaryotic cell species to grow in liquid culture on the petri dish surface without adding coatings such as ECM (extracellular matrices), proteines, collagen, fibronectin, poly-D-lysine, or specialty plasma gas treatments.