Learn More
Diversified Biotech Inc Planet-Safe 96well Round Bottom Microplate (Non Sterile)
Catalog No. NC3360900
Encompass
100% Plant-Based Planet-Safe Cell Culture Microplates are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane Superior Cell-Binding PLA is considerably more hydrophilic than petrochemical resins such as polystyrene. This beneficial property allows a variety of anchorage-dependent eukaryotic cell species to grow in liquid culture on the dish surface without added coatings or treatments.
Catalog No. NC3360900 Supplier Diversified Biotech Inc Supplier No. PLATE2000NS
Add to Cart
Add to Cart
100% Plant-Based Planet-Safe Cell Culture Microplates are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane Superior Cell-Binding PLA is considerably more hydrophilic than petrochemical resins such as polystyrene. This beneficial property allows a variety of anchorage-dependent eukaryotic cell species to grow in liquid culture on the dish surface without added coatings or treatments.