Learn More
Diversified Biotech Inc Planet-Safe Cell Culture dishes, 100mm 375/cs (Sterile)
Catalog No. NC3360885
Encompass
Planet-Safe 150 mm Cell Culture dishes are made from 100% plant-based PLA, derived from sustainably grown sugarcane. The hydrophilic PLA supports growth of anchorage-dependent cells without coatings, performing comparably to polystyrene. Eco-friendly disposal options include composting, incineration, or landfill with carbon sequestration benefits. Case of 375.
Catalog No. NC3360885 Supplier Diversified Biotech Inc Supplier No. CELL1000
Planet-Safe 150 mm Cell Culture dishes are made from 100% plant-based PLA, derived from sustainably grown sugarcane. The hydrophilic PLA supports growth of anchorage-dependent cells without coatings, performing comparably to polystyrene. Eco-friendly disposal options include composting, incineration, or landfill with carbon sequestration benefits. Case of 375.