Learn More
Diversified Biotech Inc Planet-Safe Petri Dishes, 100mm 375/cs (Sterile)
Catalog No. NC3360901
Encompass
100% Plant-Based Planet-Safe Cell Culture Petridishes are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane. Case of 375.
This item is not returnable. View return policy
Catalog No. NC3360901 Supplier Diversified Biotech Inc Supplier No. PSPD1000
100% Plant-Based Planet-Safe Cell Culture Petridishes are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane. Case of 375.