Learn More
Diversified Biotech Inc Planet-Safe Petri Dishes, 96well Flat Bottom Microplate (Non Sterile)
Catalog No. NC3360896
Encompass
100% Plant-Based Planet-Safe Cell Culture Microplates are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane Superior Cell-Binding PLA is considerably more hydrophilic than petrochemical resins such as polystyrene. This beneficial property allows a variety of anchorage-dependent eukaryotic cell species to grow in liquid culture on the dish surface without added coatings or treatments.
This item is not returnable. View return policy
Catalog No. NC3360896 Supplier Diversified Biotech Inc Supplier No. PLATE1000NS
100% Plant-Based Planet-Safe Cell Culture Microplates are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane Superior Cell-Binding PLA is considerably more hydrophilic than petrochemical resins such as polystyrene. This beneficial property allows a variety of anchorage-dependent eukaryotic cell species to grow in liquid culture on the dish surface without added coatings or treatments.