Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Planned Maintenance for Guardian 20 ft.
SDP

Catalog No. 22026915 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Guardian 20 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-026-915 Guardian 20 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-026-915 Supplier Mopec Supplier No. SV904
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Service agreements provide support to help maintain equipment operation, performance, and efficiency throughout the service period.

  • Annual preventive maintenance (PM) service includes testing, verification, and calibration of the station.
  • Service covers 14 inspection areas and more than 50 maintenance procedures.
  • Equipment is checked to ensure it meets manufacturer specifications.
  • Preventive maintenance helps identify potential issues before they result in equipment downtime.
  • Repairs and replacement parts are not included and will be quoted separately if required.

Included with the agreement:

  • Technical support available Monday to Friday, 8:00 AM - 5:00 PM.
  • One scheduled preventive maintenance visit per year.
  • Travel costs for the preventive maintenance visit are included.
  • 5% Hourly Rate Discount (non-covered events).

Specifications

For Use With (Equipment) Guardian 20 ft.
Product Type Planned Maintenance
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.