Learn More
MSC Plano Molding Compartment Box, 5 in, 1-3/4 in, 1 Compartment, Clear
Catalog No. 5000535531
Encompass
This compartment box is designed for organizing small parts and features a positive snap lock for secure closure. It measures 5 inches in width and 1-3/4 inches in depth, and has a clear design for easy content viewing.
  • Positive snap lock for secure closure
  • 5 in width and 1-3/4 in depth for compact storage
  • Single compartment for versatile use
  • Clear design allows for easy identification of contents
  • Sturdy cam-action latches ensure durability
  • Dri-Loc O-ring seal provides added protection
Catalog No. 50-005-35531 Supplier MSC Supplier No. 61065660
This compartment box is designed for organizing small parts and features a positive snap lock for secure closure. It measures 5 inches in width and 1-3/4 inches in depth, and has a clear design for easy content viewing.
  • Positive snap lock for secure closure
  • 5 in width and 1-3/4 in depth for compact storage
  • Single compartment for versatile use
  • Clear design allows for easy identification of contents
  • Sturdy cam-action latches ensure durability
  • Dri-Loc O-ring seal provides added protection