Learn More
MSC Plano Molding Storage Box, 5 in Width, 2 in Depth, Clear/Red
Catalog No. 5000529469
Encompass
This storage box is designed for organizing small parts securely. It features a positive snap lock mechanism, a clear and red design, and a single compartment. Its robust construction makes it suitable for various storage needs, ensuring contents are protected and easily accessible.
  • Positive snap lock for secure closure.
  • 5 inch width and 2 inch depth for compact storage.
  • Clear and red color combination for easy visibility of contents.
  • Includes a wrist lanyard for convenient portability.
  • Dri-Loc O-Ring Seal provides a watertight seal.
  • Sturdy cam-action latch ensures durability and reliable fastening.
  • Features a single compartment for versatile organization.
Catalog No. 50-005-29469 Supplier MSC Supplier No. 10286821
This storage box is designed for organizing small parts securely. It features a positive snap lock mechanism, a clear and red design, and a single compartment. Its robust construction makes it suitable for various storage needs, ensuring contents are protected and easily accessible.
  • Positive snap lock for secure closure.
  • 5 inch width and 2 inch depth for compact storage.
  • Clear and red color combination for easy visibility of contents.
  • Includes a wrist lanyard for convenient portability.
  • Dri-Loc O-Ring Seal provides a watertight seal.
  • Sturdy cam-action latch ensures durability and reliable fastening.
  • Features a single compartment for versatile organization.