Learn More
3B Scientific™ Plant Cell Slide Set
SDPSCI_ED
Catalog No. S95063L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
12
Catalog No. Quantity
S95063L 12
1 options
Catalog No. S95063L Supplier 3B Scientific™ Supplier No. W13053
Add to Cart
Add to Cart
12 microscope slides
  • 1(c). Epidermis of Allium (onion), w.m. showing simple plant cells with cell walls, nuclei and
  • cytoplasm
  • 2(d). Root tips of Allium cepa l.s. showing cell division (mitosis) in all stages
  • 3(e). Pollen mother cells of Lilium. Prophase of first maturation division (meiosis)
  • 4(f). Pollen mother cells of Lilium. Metaphase and anaphase of first maturation division
  • 5(c). Wood of Tilia macerated and w.m.
  • 6(d). Fruit of Pyrus (pear) t.s. showing stone cells
  • 7(c). Tuber of Solanum (potato) t.s. shows cork and starch grains
  • 8(d). Cucurbita pepo (pumpkin) l.s. of stem showing vascular bundles with sieve tubes, spiral and
  • annular vessels
  • 9(c). Ricinus endosperm t.s. showing aleurone grains
  • 10(d). Anthers of Lilium (lily), t.s. pollen sacs and pollen grains
  • 11(d). Ovary of Lilium (lily), t.s. arrangement of ovules and embryosac
  • 12(e). Spirogyra showing conjugation stages and zygotes
Specifications
Type Plant Cell
Quantity 12