Learn More
Research Products International Corp Plant Culture Vessel, 65mm x 65mm x 77mm, 100 Vessels per Case
SDP
Catalog No. 50998443
Encompass
High quality plant culture vessels molded from durable polycarbonate and provide high relative growing area for plants.
Can be autoclaved for repeated use.
Supplied with lids.
Provides convenient handling and storage with uniform lighting.
Catalog No. 50-998-443 Supplier Research Products International Corp Supplier No. 248950
High quality plant culture vessels molded from durable polycarbonate and provide high relative growing area for plants.
Can be autoclaved for repeated use.
Supplied with lids.
Provides convenient handling and storage with uniform lighting.