Learn More
Bioworld PLANT/FUNGI DNA ISOLATION KIT
Catalog No. 50196971
Encompass_Preferred
For rapid isolation of total DNA from plants and fungi Rapid and simple procedure; Isolate total DNA including pathogen DNA without phenol; Liquid nitrogen is optional for homogenization; Available in spin column format adn 96-well form...
Catalog No. 50-196-971 Supplier Bioworld Supplier No. 107601141
For rapid isolation of total DNA from plants and fungi Rapid and simple procedure; Isolate total DNA including pathogen DNA without phenol; Liquid nitrogen is optional for homogenization; Available in spin column format adn 96-well form...