Learn More
Epoch Life Science Inc GenCatch™ Plant Genomic DNA Purification System (250 preps) Uniquely formulated buffers, shearing tubes, reinforced columns with collection tubes, along with protocols.
SDP
Supplier:  Epoch Life Science Inc 1560250
Encompass_Preferred
Provides a simple, fast and cost-effective method to extract gDNA ready for downstream use from various plant species. Comes with shearing tubes for simple and fast homogenization and filtration of plant tissues. 40 ug yield. 100 mg plant tissue. 200 ul elution volume. Designed predominantly for gDNA of 20-30 kb. No phenol/chloroform extraction nor ethanol precipitation required. Used by labs around the world. 18 month shelf life.
Catalog No. 50-153-8095