Learn More
Percival Scientific Plant Growth Chamber, E30L1C9, 9.7 cu. ft., SciWhite with red LEDs, 1 tier, 7-44° C ±0.5°, IntellusUltraConnect controller, touchscreen interface, remote connectivity
SDP
Catalog No. NC3794976
Encompass_Preferred
The E30L1C9 plant growth chamber is frequently used for Alfalfa/forages, peanut, potato, bluegrass, short term plant growth and/or short plants. SciWhite w/ red LEDs are dimmable between 10-100% output with up to 870 µmoles/m2/s light irradiance @ 6”. IntellusUltraConnect controller, touchscreen interface, remote connectivity. Interior/exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and stainless-steel floor for corrosion resistance. One white epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Rear chamber wall mounted evaporator coil incorporates an air circulation fan. For additional options contact sales@percival-scientific.com for a free consultation.
Catalog No. NC3794976 Supplier Percival Scientific Supplier No. E30L1C9
Add to Cart
Add to Cart
Request a Quote
The E30L1C9 plant growth chamber is frequently used for Alfalfa/forages, peanut, potato, bluegrass, short term plant growth and/or short plants. SciWhite w/ red LEDs are dimmable between 10-100% output with up to 870 µmoles/m2/s light irradiance @ 6”. IntellusUltraConnect controller, touchscreen interface, remote connectivity. Interior/exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and stainless-steel floor for corrosion resistance. One white epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Rear chamber wall mounted evaporator coil incorporates an air circulation fan. For additional options contact sales@percival-scientific.com for a free consultation.