Learn More
Percival Scientific Plant Growth Chamber, E36L1, 29.7 cu. ft., SciWhite with red LEDs, 1 tier, 7-44° C ±0.5°, IntellusUltra controller, touchscreen interface
SDP
Catalog No. NC3794982
Encompass_Preferred
The E36L1 growth chamber is frequently used for soybean, rice, tomato, cotton or other short to medium height plants. SciWhite with red LEDs are dimmable from 10-100% output with up to 680 µmoles/m2/s irradiance @ 6". IntellusUltra controller, touchscreen interface. Interior/ exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. One white epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.
Catalog No. NC3794982 Supplier Percival Scientific Supplier No. E36L1
Add to Cart
Add to Cart
Request a Quote
The E36L1 growth chamber is frequently used for soybean, rice, tomato, cotton or other short to medium height plants. SciWhite with red LEDs are dimmable from 10-100% output with up to 680 µmoles/m2/s irradiance @ 6". IntellusUltra controller, touchscreen interface. Interior/ exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. One white epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.