Learn More
Percival Scientific Plant Growth Chamber, E36L1H14, 29.7 cu. ft., SciWhite with red LEDs, 1 tier,, 7-44° C ±0.5°, IntellusUltra controller, touchscreen, standard additive ultrasonic humidifier
SDP
Catalog No. NC3795067
Encompass_Preferred
The E36L1H14 growth chamber is frequently used for rice, cotton or short/medium height plants. SciWhite with red LEDs are dimmable from 10-100% output w/ up to 680 µmoles/m2/s irradiance @ 6". IntellusUltra controller, touchscreen, standard additive ultrasonic humidifier. Interior/exterior of smooth white galvanized steel w/ foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. One white epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit w/ hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.
Catalog No. NC3795067 Supplier Percival Scientific Supplier No. E36L1H14
Add to Cart
Add to Cart
Request a Quote
The E36L1H14 growth chamber is frequently used for rice, cotton or short/medium height plants. SciWhite with red LEDs are dimmable from 10-100% output w/ up to 680 µmoles/m2/s irradiance @ 6". IntellusUltra controller, touchscreen, standard additive ultrasonic humidifier. Interior/exterior of smooth white galvanized steel w/ foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. One white epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit w/ hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.