Learn More
Percival Scientific Plant Growth Chamber, E41L1C9, 37.2 cu. ft., SciWhite with red LEDs, 1 tier, 7-44° C ±0.5°, IntellusUltraConnect controller, touchscreen interface, remote connectivity
SDP
Catalog No. NC3795097
Encompass_Preferred
The E41L1C9 growth chamber is frequently used for soybean, tomato, cotton or other short/medium height plants. SciWhite with red LEDs are dimmable from 10-100% output with up to 800 µmoles/m2/s irradiance @ 6". IntellusUltraConnect controller, touchscreen, remote connectivity. Interior/exterior of smooth white galvanized steel w/ foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. One epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.
Catalog No. NC3795097 Supplier Percival Scientific Supplier No. E41L1C9
Add to Cart
Add to Cart
Request a Quote
The E41L1C9 growth chamber is frequently used for soybean, tomato, cotton or other short/medium height plants. SciWhite with red LEDs are dimmable from 10-100% output with up to 800 µmoles/m2/s irradiance @ 6". IntellusUltraConnect controller, touchscreen, remote connectivity. Interior/exterior of smooth white galvanized steel w/ foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. One epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.