Learn More
Percival Scientific Plant Growth Chamber, E75L1C9, 76.1 cu. ft., SciWhite with red LEDs, 1 tier, 7-44° C ±0.5°, IntellusUltraConnect controller, touchscreen interface, remote connectivity
SDP
Catalog No. NC3795583
Encompass_Preferred
The E75L1C9 growth chamber is frequently used for cereals, citrus, grasses and other plants that require high light intensity and higher growth height. SciWhite with red LEDs are dimmable from 10-100% output with up to 1360 µmoles/m2/s irradiance @ 6". IntellusUltraConnect controller, touchscreen interface, remote connectivity. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. One white epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Contact sales@percival-scientific.com for more options.
Catalog No. NC3795583 Supplier Percival Scientific Supplier No. E75L1C9
Add to Cart
Add to Cart
Request a Quote
The E75L1C9 growth chamber is frequently used for cereals, citrus, grasses and other plants that require high light intensity and higher growth height. SciWhite with red LEDs are dimmable from 10-100% output with up to 1360 µmoles/m2/s irradiance @ 6". IntellusUltraConnect controller, touchscreen interface, remote connectivity. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. One white epoxy coated steel wire shelf adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Contact sales@percival-scientific.com for more options.