Learn More
Percival Scientific Plant Growth Chamber, PGC105C9, 108.3 cu. ft., SciWhite with red LEDs, 1 tier, 7-44° C ±0.5°, Upgraded IntellusUltraConnect controller, touchscreen interface, remote connectivity
SDP
Catalog No. NC3795597
Encompass_Preferred
The PGC105C9 growth chamber is frequently used for cereals, citrus, grasses and other plants that require high light intensity and higher growth height. SciWhite with red LED lampbank is vertically adjustable and dimmable from 10-100% output with up to 1450 µmoles/m2/s irradiance @ 6". Upgraded IntellusUltraConnect control system. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Conditioned air moves in uniform upward direction through entire work bench through perforations in aluminum channels. Fresh air inlet and outlet are adjustable. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Contact sales@percival-scientific.com for more options.
Catalog No. NC3795597 Supplier Percival Scientific Supplier No. PGC105C9
Add to Cart
Add to Cart
Request a Quote
The PGC105C9 growth chamber is frequently used for cereals, citrus, grasses and other plants that require high light intensity and higher growth height. SciWhite with red LED lampbank is vertically adjustable and dimmable from 10-100% output with up to 1450 µmoles/m2/s irradiance @ 6". Upgraded IntellusUltraConnect control system. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Conditioned air moves in uniform upward direction through entire work bench through perforations in aluminum channels. Fresh air inlet and outlet are adjustable. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Contact sales@percival-scientific.com for more options.