Learn More
Percival Scientific PERCIVAL E-36L2 Plant Growth Chamber, IntellusUltra Controller, SciWhite LED Lighting System, Unit
SDP
Catalog No. NC2891316
Encompass_Preferred
The PERCIVAL SCIENTIFIC E-36L2 Plant Growth Chamber is expertly designed for plant research, providing precise environmental control essential for vascular plant development, seedling germination, and more. With its advanced IntellusUltra controller and a SciWhite LED lighting system, it ensures optimal growth conditions with customizable light output and efficient airflow. This chamber stands out for its precise management capabilities and robust construction, making it a reliable choice for various scientific applications.
Catalog No. NC2891316 Supplier Percival Scientific Supplier No. E36L2
Add to Cart
Add to Cart
Request a Quote
The PERCIVAL SCIENTIFIC E-36L2 Plant Growth Chamber is expertly designed for plant research, providing precise environmental control essential for vascular plant development, seedling germination, and more. With its advanced IntellusUltra controller and a SciWhite LED lighting system, it ensures optimal growth conditions with customizable light output and efficient airflow. This chamber stands out for its precise management capabilities and robust construction, making it a reliable choice for various scientific applications.