Learn More
American Research Products Inc Plant hormone abscisic acid A
Supplier:  American Research Products Inc CSBE09159PL
Encompass
Plant hormone abscisic acid ABA ELISA Kit from CUSABIO Detection Range 0 156 g/ml-10 g/ml Sensitivity 0 04 g/mL Sample plant tissues Method Competitive
Catalog No. 50-003-28656