Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plant Kingdom Poster
SCI_ED

Catalog No. S01955L Shop All Fischer Technical Company Products
Change view
Click to view available options
Format:
Poster
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S01955L Poster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S01955L Supplier Fischer Technical Company Supplier No. A112L

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The revolutionary changes in the way we look at animal life have had little impact on the plant kingdom.

  • Matches the graphic design of Animal Kingdom 2
  • Added the four divisions of extinct plants

Specifications

Format Poster
Product Type Plant Kingdom Poster
For Use With (Application) Detailed depictions and images of the Linnaeus classifications of the Plant Kingdom
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.