Learn More
LaMotte™ Plant Macronutrient Kit
SDPSCI_ED
Catalog No. S85041 Shop All LaMotte Company Products
Change view
Click to view available options
Class Size:
1 Class
Catalog No. Class Size
S85041 1 Class
1 options
This item is not returnable. View return policy
Catalog No. S85041 Supplier LaMotte™ Supplier No. 502601
Add to Cart
Add to Cart
This item is not returnable. View return policy
Perform colorimetric tests for various factors in plant tissue liquid extracts.
  • Offers tests for nitrogen, phosphorus and potassium
  • Quantitative results given as abundant, adequate and deficient only
  • Sufficient reagents for 50 tests per factor
Specifications
For Use With (Application) Provides essential information to determine nitrates, phosphorus and potassium in plant tissue concerning plant use of nutrients vital to their growth
Includes 2 x 120mL Concentrated universal extracting solution, 30mL Nitrate Reagent, 30g Nitrate reagent powder, 30mL Phosphorus reagent, 50 Phosphorus reagent tablets, 50 Potassium reagent B tablets, 120mL Potassium reagent C, 1 Plastic funnel, 1 Filter papers (100 sheets), 1 Potash A test tube with cap, 1 Phosphorus B test tube, 7 and 14mL 2 extraction test tubes with caps, 1 Plastic spot plate, 0.5g spoon, 1 Plastic stirring rod, 1mL 2 Plastic pipets, Plastic transfer pipet, Plain Glass and Plastic pipet with cap, 1 LaMotte™ soil handbook
Class Size 1 Class
Product Type Macronutrient Plant Tissue Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov