Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Plant Nutrition Chemical Package
SDPSCI_ED

Catalog No. S18439A Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S18439A 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S18439A Supplier LaMotte™ Supplier No. 5940
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A study of plant nutrition the whole class will enjoy.

Using hydroponics, control nutrients available to plants by varying the solution in which they are cultivated.

  • Package contains 10 stock solutions of macronutrients and trace elements
  • Solutions are used to make up the complete plant nutrient mixture, and different plant nutrient mixtures that are minus Ca, Mg, K, N, P, S, Fe and trace elements
  • Each solution makes 5 L of each of the mixtures
  • Instructions for use contained in Dr. Robert W. Stegner laboratory manual, Plant Nutrition Studies

Order Info

Grades 6+.

Specifications

Grade 6
For Use With (Application) Hydroponics' procedures and investigations
Includes 250ml 1m calcium nitrate, 120ml 1m magnesium sulfate, 250ml 1m potassium nitrate, 60ml 1m potassium phosphate, 120ml 0.05m calcium phosphate, 60ml 0.5m potassium sulfate, 60ml 1m magnesium nitrate, (4) 250ml bottles containing 0.01m calcium sulfate, 60ml
Product Type Plant Nutrition Studies Chemical Package
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.