Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plant Pigments and Photosynthesis Lab
SCI_ED

Catalog No. S32625
Change view
Click to view available options
Plant Pigment & Photosynthesis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S32625 Plant Pigment & Photosynthesis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S32625 Supplier Neo/Sci Corporation Supplier No. 201393
This item is not returnable. View return policy

Separate plant pigments using paper chromatography.

Masked by green chlorophyll in the summer, the abundance of pigments in plant leaves becomes highly conspicuous each fall, as differences in the types and amounts of the pigments provide visible color variations. Separate these plant pigments using a technique called paper chromatography. Pigments migrate on a specially treated paper. Each pigment's migration rate depends on its solubility and degree of attraction to the paper.

  • Uses a safer alcohol solution as an alternative to acetone
  • Safely separate plant pigments, identify them by color and position on the chromatograph and relate them to their role in photosynthesis

Order Info

Class size: 40 students.

Specifications

For Use With (Application) Study plant chromatography by separate plant pigments using paper chromatography
Requires Safer alcohol solution
Class Size 40 Students
Product Type Plant Pigments and Photosynthesis Lab Investigation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.