Learn More
PLANT CELL TECHNOLOGY Broad-spectrum biocide for plant tissue culture, in-vitro laboratory use only, Plant Preservative Mixture™, PPM™, Clear Liquid, Anti-Contamination, 500 mL Bottle
Catalog No. NC2790487
Encompass_Preferred
Plant Preservative Mixture™ (PPM™) is a liquid additive designed to reduce microbial contamination in plant tissue culture and micropropagation applications. This 500mL bottle provides ready-to-use solution that integrates easily into standard culture media. PPM™ contains broad-spectrum antimicrobial and antifungal components compatible with a wide variety of plant species, including ornamentals, woody plants, and edible crops. It supports aseptic culture and enhances explant survival rates in tissue culture applications. Packaged in a durable, clear 500mL bottle with secure cap, PPM™ ensures safe handling and storage. Ideal for research laboratories, commercial propagation, and specialized plant cell culture applications. Stable under recommended storage conditions.
Catalog No. NC2790487 Supplier PLANT CELL TECHNOLOGY Supplier No. PPM500
Plant Preservative Mixture™ (PPM™) is a liquid additive designed to reduce microbial contamination in plant tissue culture and micropropagation applications. This 500mL bottle provides ready-to-use solution that integrates easily into standard culture media. PPM™ contains broad-spectrum antimicrobial and antifungal components compatible with a wide variety of plant species, including ornamentals, woody plants, and edible crops. It supports aseptic culture and enhances explant survival rates in tissue culture applications. Packaged in a durable, clear 500mL bottle with secure cap, PPM™ ensures safe handling and storage. Ideal for research laboratories, commercial propagation, and specialized plant cell culture applications. Stable under recommended storage conditions.