Learn More
PLANT CELL TECHNOLOGY Plant Preservative Mixture (PPM™), Broad-spectrum biocide/fungicide, 0.05% - 0.2% v/v, Heat stable at 121°C, Completely soluble in water, Storage at 4°C
Supplier:  PLANT CELL TECHNOLOGY 1000
Encompass
The Plant Preservative Mixture (PPM™) from Plant Cell Technology is a heat-stable biocide and fungicide formulated to control microbial contamination in *in vitro* plant tissue cultures. It effectively prevents the germination and growth of harmful microorganisms while preserving critical plant development processes, making it a cost-effective alternative to traditional antibiotics. PPM™ simplifies integration into media preparation protocols and enhances the success rates of culture experiments by managing various types of contamination.
  • Broad-spectrum efficacy against bacteria, fungi, yeasts, and molds
  • Heat stable; effective after autoclaving at 121°C for 20 min
  • Non-phytotoxic at recommended doses
  • Reduces airborne, waterborne, and human-introduced contamination
  • Economically priced compared to conventional antibiotics
  • Targets key microbial enzymatic pathways to reduce resistance
Catalog No. NC0412973